ENA402268117 Enamine

butane-2-sulfonamide

  • MDL number MFCD12783445
  • Cas Number : 17854-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75253

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75253
Availability details for
Quantity
  • Availability
  • Add to Cart