ENA402268144 Enamine

8-oxatricyclo[7.4.0.0^{2,7}]trideca-1(9),2(7),3,5,10,12-hexaen-5-ol

  • MDL number MFCD00094159
  • Cas Number : 20279-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91339

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91339
Availability details for
Quantity
  • Availability
  • Add to Cart