ENA402268148 Enamine

3-methylbutane-2-sulfonyl chloride

  • MDL number MFCD16671792
  • Cas Number : 78429-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95198

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95198
Availability details for
Quantity
  • Availability
  • Add to Cart