ENA402268151 Enamine

methyl 3-hydroxybutanoate

  • MDL number MFCD00129999
  • Cas Number : 1487-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Flash Point 72
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95930

Safety Information

signalword Warning
safety_statement 23-24/25
msds https://www.enaminestore.com/catalog/EN300-95930
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart