ENA405193840 Enamine

methyl 3-(1-cyanocyclobutyl)butanoate

  • MDL number MFCD25970259
  • Cas Number : 1461713-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124286
Availability details for
Quantity
  • Availability
  • Add to Cart