ENA405193842 Enamine

5-bromo-8-(bromomethyl)quinoline

  • MDL number MFCD25970263
  • Cas Number : 88474-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-112
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127822
Availability details for
Quantity
  • Availability
  • Add to Cart