ENA405193843 Enamine

(2S)-2,3-diaminopropan-1-ol dihydrochloride

  • MDL number MFCD25970264
  • Cas Number : 87584-94-9
  • Molecular Weight : 163.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 193-195
Iupac Name (S)-2,3-diaminopropan-1-ol dihydrochloride
Inchi Code 1S/C3H10N2O.2ClH/c4-1-3(5)2-6;;/h3,6H,1-2,4-5H2;2*1H/t3-;;/m0../s1
Inchi Key IKBUWJHUBADHSZ-QTNFYWBSSA-N

Description

Melting Point 193-195
Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-128518

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-128518
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart