ENA405193849 Enamine

[4-methyl-5-(methylsulfanyl)-4H-1,2,4-triazol-3-yl]methanamine dihydrochloride

  • MDL number MFCD25970272
  • Cas Number : 1461706-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129427

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129427
Availability details for
Quantity
  • Availability
  • Add to Cart