ENA405193855 Enamine

methyl 2-[(2,4-dichlorophenyl)formamido]-3-hydroxybutanoate

  • MDL number MFCD25970280
  • Cas Number : 1485648-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130079
Availability details for
Quantity
  • Availability
  • Add to Cart