ENA405193857 Enamine

2-(2-methoxyethyl)-4,5,6,7-tetrahydro-2H-indazol-7-amine

  • MDL number MFCD25970282
  • Cas Number : 1461707-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130408
Availability details for
Quantity
  • Availability
  • Add to Cart