ENA405193858 Enamine

methyl 3-({[amino(4-bromophenyl)methylidene]amino}oxy)prop-2-enoate

  • Cas Number : 1461727-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130852
Availability details for
Quantity
  • Availability
  • Add to Cart