ENA405193863 Enamine

methyl 2-(2-methoxyethyl)-4,5-dihydro-1,3-oxazole-4-carboxylate

  • MDL number MFCD25970293
  • Cas Number : 1461706-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131571

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131571
Availability details for
Quantity
  • Availability
  • Add to Cart