ENA405193867 Enamine

2-(5-chloro-2,3-dihydro-1-benzofuran-7-yl)acetonitrile

  • MDL number MFCD25970299
  • Cas Number : 1461707-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100...102
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131795

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131795
Availability details for
Quantity
  • Availability
  • Add to Cart