ENA405193868 Enamine

ethyl 5-ethoxy-1-[2-(trifluoromethyl)phenyl]-4,5-dihydro-1H-pyrazole-3-carboxylate

  • MDL number MFCD25970300
  • Cas Number : 1461714-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131863
Availability details for
Quantity
  • Availability
  • Add to Cart