ENA405193869 Enamine

methyl 2,6-dimethylpyrimidine-4-carboxylate

  • MDL number MFCD25970303
  • Cas Number : 54198-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55...57
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132010

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132010
Availability details for
Quantity
  • Availability
  • Add to Cart