ENA405193876 Enamine

2-[(2-hydroxy-2-methylpropyl)amino]-4-nitropyridin-1-ium-1-olate

  • MDL number MFCD25970312
  • Cas Number : 1461715-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132803
Availability details for
Quantity
  • Availability
  • Add to Cart