ENA405193877 Enamine

3-chloro-4-(dibromomethyl)benzoic acid

  • MDL number MFCD25970313
  • Cas Number : 1403326-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-132810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132810
Availability details for
Quantity
  • Availability
  • Add to Cart