ENA405193879 Enamine

1-(4,5-dicyclopropyl-4H-1,2,4-triazol-3-yl)methanamine dihydrochloride

  • MDL number MFCD25970315
  • Cas Number : 1461715-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132897
Availability details for
Quantity
  • Availability
  • Add to Cart