ENA405193882 Enamine

4-(3-methoxypropoxy)-3-nitrobenzonitrile

  • MDL number MFCD25970320
  • Cas Number : 1461715-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87...89
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133155

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133155
Availability details for
Quantity
  • Availability
  • Add to Cart