ENA405193887 Enamine

5-chloro-2,3-dihydro-1-benzofuran-7-sulfonyl chloride

  • MDL number MFCD25970326
  • Cas Number : 1461708-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133214

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133214
Availability details for
Quantity
  • Availability
  • Add to Cart