ENA405193889 Enamine

1-acetylpiperidine-4-carboximidamide hydrochloride

  • MDL number MFCD25970330
  • Cas Number : 1461713-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133235
Availability details for
Quantity
  • Availability
  • Add to Cart