ENA405193892 Enamine

2-(2-chloro-6-fluorophenyl)-2-(piperidin-1-yl)acetic acid hydrochloride

  • MDL number MFCD25970334
  • Cas Number : 1461704-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...212
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133239

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133239
Availability details for
Quantity
  • Availability
  • Add to Cart