ENA405193894 Enamine

3,5,5-trimethylpiperidin-2-one

  • MDL number MFCD25970338
  • Cas Number : 1461709-34-1
  • Molecular Weight : 141.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,5,5-trimethylpiperidin-2-one
Inchi Code 1S/C8H15NO/c1-6-4-8(2,3)5-9-7(6)10/h6H,4-5H2,1-3H3,(H,9,10)
Inchi Key CZDWYMQBLSPGKI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-133285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133285
Availability details for
Quantity
  • Availability
  • Add to Cart