ENA405193897 Enamine

2-(2H-1,3-benzodioxol-5-yl)-N-methoxy-N-methylacetamide

  • MDL number MFCD25959404
  • Cas Number : 153277-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133305

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133305
Availability details for
Quantity
  • Availability
  • Add to Cart