ENA405193899 Enamine

tert-butyl N-(4,5,6,7-tetrahydro-1H-indazol-7-yl)carbamate

  • MDL number MFCD25970343
  • Cas Number : 1461708-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133369

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133369
Availability details for
Quantity
  • Availability
  • Add to Cart