ENA405193903 Enamine

benzyl 3-[4-(2-hydroxypropan-2-yl)-1H-1,2,3-triazol-1-yl]azetidine-1-carboxylate

  • MDL number MFCD25970348
  • Cas Number : 1461715-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125...127
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133402

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133402
Availability details for
Quantity
  • Availability
  • Add to Cart