ENA405193906 Enamine

methyl 2-[cyclopropyl(2,3-difluoro-6-nitrophenyl)amino]acetate

  • MDL number MFCD25970354
  • Cas Number : 1461708-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133462
Availability details for
Quantity
  • Availability
  • Add to Cart