ENA405193908 Enamine

1-(1-benzoyl-4-methylpiperidine-4-carbonyl)-4-methylpiperazine

  • MDL number MFCD25970356
  • Cas Number : 1461709-14-7
  • Molecular Weight : 329.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133471

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133471
Availability details for
Quantity
  • Availability
  • Add to Cart