ENA405193909 Enamine

4-[(4-ethylphenyl)methyl]piperidine-4-carboxylic acid hydrochloride

  • MDL number MFCD25970357
  • Cas Number : 1461714-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133477
Availability details for
Quantity
  • Availability
  • Add to Cart