ENA405193910 Enamine

3-ethyl-1-(pyrrolidin-3-yl)urea hydrochloride

  • MDL number MFCD25970358
  • Cas Number : 1461709-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133491
Availability details for
Quantity
  • Availability
  • Add to Cart