ENA405193913 Enamine

4-bromo-2,6-dimethyl-N-[(1,2-oxazol-4-yl)methyl]aniline hydrobromide

  • MDL number MFCD25970362
  • Cas Number : 1461713-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133508
Availability details for
Quantity
  • Availability
  • Add to Cart