ENA405193914 Enamine

2-methoxy-1-(4-nitro-2,3-dihydro-1H-indol-1-yl)ethan-1-one

  • MDL number MFCD25970363
  • Cas Number : 1461713-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133519
Availability details for
Quantity
  • Availability
  • Add to Cart