ENA405193918 Enamine

tert-butyl N-[2-(4-methyl-5-phenyl-4H-1,2,4-triazol-3-yl)ethyl]carbamate

  • MDL number MFCD25970369
  • Cas Number : 1461715-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133686

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133686
Availability details for
Quantity
  • Availability
  • Add to Cart