ENA405193921 Enamine

1,3-diethyl 2-(2-fluoro-4-nitrophenyl)-2-methylpropanedioate

  • MDL number MFCD25970372
  • Cas Number : 88975-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133709

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133709
Availability details for
Quantity
  • Availability
  • Add to Cart