ENA405193925 Enamine

sodium 2-fluoro-2-(4-methyl-1,3-thiazol-2-yl)acetate

  • MDL number MFCD25970376
  • Cas Number : 1461713-66-5
  • Molecular Weight : 197.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-133721

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133721
Availability details for
Quantity
  • Availability
  • Add to Cart