ENA405193926 Enamine

{3-fluoro-1-[(1R)-1-phenylethyl]pyrrolidin-3-yl}methanol

  • MDL number MFCD25970377
  • Cas Number : 1461698-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133753
Availability details for
Quantity
  • Availability
  • Add to Cart