ENA405193932 Enamine

tert-butyl 3-[5-(methoxymethyl)-1,2,4-oxadiazol-3-yl]morpholine-4-carboxylate

  • MDL number MFCD25970385
  • Cas Number : 1461708-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133845
Availability details for
Quantity
  • Availability
  • Add to Cart