ENA405193934 Enamine

lithium(1+) 5-ethyl-1,3-thiazole-2-carboxylate

  • MDL number MFCD25970387
  • Cas Number : 1461709-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133849
Availability details for
Quantity
  • Availability
  • Add to Cart