ENA405193937 Enamine

1-[2-amino-3-(4-methylbenzoyl)-4H,5H,6H,7H-thieno[2,3-c]pyridin-6-yl]ethan-1-one

  • MDL number MFCD25970390
  • Cas Number : 1256257-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133871

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133871
Availability details for
Quantity
  • Availability
  • Add to Cart