ENA405193942 Enamine

4-(2-methylphenyl)-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD25970398
  • Cas Number : 83674-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240...242
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133944

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133944
Availability details for
Quantity
  • Availability
  • Add to Cart