ENA405193943 Enamine

4-methyl-1-(pyrrolidine-3-carbonyl)piperidine hydrochloride

  • MDL number MFCD25970399
  • Cas Number : 1461713-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133962
Availability details for
Quantity
  • Availability
  • Add to Cart