ENA405193948 Enamine

N-(3-aminopropyl)-2-fluoroaniline hydrochloride

  • MDL number MFCD25970405
  • Cas Number : 1461714-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176...178
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134054

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134054
Availability details for
Quantity
  • Availability
  • Add to Cart