ENA405193951 Enamine

{imidazo[1,2-a]pyridin-3-ylmethyl}dimethylamine hydrochloride

  • MDL number MFCD25970411
  • Cas Number : 1461708-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134071

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134071
Availability details for
Quantity
  • Availability
  • Add to Cart