ENA405193953 Enamine

2-(1,4-diazepan-1-yl)-5-nitro-1H-1,3-benzodiazole dihydrochloride

  • MDL number MFCD25970414
  • Cas Number : 1461707-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 290...292
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134131

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134131
Availability details for
Quantity
  • Availability
  • Add to Cart