ENA405193954 Enamine

1-(2,5-dichlorophenyl)-1-phenylmethanamine hydrochloride

  • MDL number MFCD25970415
  • Cas Number : 1461706-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134147
Availability details for
Quantity
  • Availability
  • Add to Cart