ENA405193955 Enamine

2-(prop-2-yn-1-yloxy)ethan-1-amine hydrochloride

  • MDL number MFCD25970416
  • Cas Number : 1383604-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134148
Availability details for
Quantity
  • Availability
  • Add to Cart