ENA405193956 Enamine

5-oxo-[1,3'-bipyrrolidine]-3-carboxylic acid hydrochloride

  • MDL number MFCD25970417
  • Cas Number : 1461714-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134149

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134149
Availability details for
Quantity
  • Availability
  • Add to Cart