ENA405193957 Enamine

3-(1H-imidazol-1-yl)-1-(4-methoxyphenyl)propan-1-one

  • MDL number MFCD25970418
  • Cas Number : 176645-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134194

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134194
Availability details for
Quantity
  • Availability
  • Add to Cart