ENA405193958 Enamine

3-(1H-imidazol-1-yl)-1-(4-methylphenyl)propan-1-one

  • MDL number MFCD25970419
  • Cas Number : 405213-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63...65
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134200

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134200
Availability details for
Quantity
  • Availability
  • Add to Cart