ENA405193964 Enamine

3-methyl-5-phenyl-1,2-oxazol-4-amine hydrochloride

  • MDL number MFCD25970426
  • Cas Number : 1461714-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134275

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134275
Availability details for
Quantity
  • Availability
  • Add to Cart